accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 138377-34-1

OHClFFF

CAS RN 138377-34-1

IUPAC Name: 2-chloro-3-(trifluoromethyl)phenol
Synonyms: 2-chloro-3-(trifluoromethyl)phenol
Molecular Weight (g/mol): 196.55
Molecular Formula: C7H4ClF3O
InChi Key: BGXZMBZFGAIBDH-UHFFFAOYSA-N
SMILES: OC1=CC=CC(=C1Cl)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products