accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 138526-69-9

FBrFF

CAS RN 138526-69-9

IUPAC Name: 5-bromo-1,2,3-trifluorobenzene
Synonyms: 1-bromo-3,4,5-trifluorobenzene
Molecular Weight (g/mol): 210.98
Molecular Formula: C6H2BrF3
InChi Key: HKJCELUUIFFSIN-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC(F)=C1F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products