accessibility menu, dialog, popup

UserName

CAS RN 13861-22-8

H3CCH2OOCH

CAS RN 13861-22-8

IUPAC Name: prop-2-yn-1-yl 2-methylprop-2-enoate
Synonyms: prop-2-yn-1-yl 2-methylprop-2-enoate
Molecular Weight (g/mol): 124.14
Molecular Formula: C7H8O2
InChi Key: PZAWASVJOPLHCJ-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCC#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results