CAS RN 13874-04-9
CAS RN 13874-04-9
IUPAC Namn:
ammonium tetrachlorogolduide hydrate
Synonymer:
ammonium hydrate tetrachloroaurate
Molekylvikt (g/mol):
374.82
Molekylär sammansättning:
AuCl4H6NO
InChi Key:
DTYXSEJVOCMIPK-UHFFFAOYSA-K
SMILES:
[NH4+].O.Cl[Au-](Cl)(Cl)Cl
1
–
1
av
1
Resultat
1
Ammonium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis)
CAS: 13874-04-9 Molekylformel: AuCl4H4N Molekylvikt (g/mol): 356.82 MDL-nummer: MFCD00011503 InChI-nyckel: APXMUHKEDBIQKQ-UHFFFAOYSA-J Synonym: ammonium tetrachloroaurate iii hydrate,ammonium tetrachloroaurate iii hydrate, premion,ammonium tetrachloroaurate ii,azanium;tetrachlorogold 1-;hydrate,ammonium tetrachloroaurate iii n-hydrate,ammonium tetrachloroaurate 1--water 1/1/1,ammoniumtetrachloroaurate iii hydrate-au puratrem,ammonium tetrachloroaurate iii hydrate, premion r metals basis PubChem CID: 16211474 LEDER: N.Cl[Au](Cl)(Cl)Cl