accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 139-40-2

CH3CH3HNNClNNHH3CCH3N

CAS RN 139-40-2

IUPAC Name: 6-chloro-N2,N4-bis(propan-2-yl)-1,3,5-triazine-2,4-diamine
Synonyms: propazine prozinex
Molecular Weight (g/mol): 229.71
Molecular Formula: C9H16ClN5
InChi Key: WJNRPILHGGKWCK-UHFFFAOYSA-N
SMILES: CC(C)NC1=NC(NC(C)C)=NC(Cl)=N1
Solvent or Matrix 
  • (2)
  • (2)
  • (2)
  • (1)
Concentration 
  • (3)
  • (3)

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products