man

Test

CAS RN 139177-63-2

FFFSOOOFFFSOOOFFFSOOOHo

CAS RN 139177-63-2

IUPAC Name: holmium(3+) tritrifluoromethanesulfonate
Synonyms: holmium(3+) tritriflate
Molecular Weight (g/mol): 612.12
Molecular Formula: C3F9HoO9S3
InChi Key: DBPDCYACEYLEMY-UHFFFAOYSA-K
SMILES: [Ho+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results