CAS RN 13925-07-0
CAS RN 13925-07-0
IUPAC Name:
2-ethyl-3,5-dimethylpyrazine
Synonyms:
2-ethyl-3,5-dimethylpyrazine
Molecular Weight (g/mol):
136.2
Molecular Formula:
C8H12N2
InChi Key:
JZBCTZLGKSYRSF-UHFFFAOYSA-N
SMILES:
CCC1=NC=C(C)N=C1C
1
–
1
of
1
results
1
2-Ethyl-3,5(6)-dimethylpyrazine, 99%, mixture of isomers, Thermo Scientific Chemicals
CAS: 13925-07-0 Molecular Formula: C8H12N2 Molecular Weight (g/mol): 136.198 MDL Number: MFCD00047392 InChI Key: JZBCTZLGKSYRSF-UHFFFAOYSA-N Synonym: 3,5-dimethyl-2-ethylpyrazine,pyrazine, 2-ethyl-3,5-dimethyl,2,6-dimethyl-3-ethylpyrazine,3-ethyl-2,6-dimethylpyrazine,2-ethyl-3,5-dimethyl pyrazine,pyrazine, 2,6-dimethyl-3-ethyl,unii-r182ey5l8c,fema no. 3149,2-ethyl-3,5 6-dimethylpyrazine,pyrazine, 3,5-dimethyl-2-ethyl PubChem CID: 26334 IUPAC Name: 2-ethyl-3,5-dimethylpyrazine SMILES: CCC1=NC=C(N=C1C)C