CAS RN 139512-70-2
CAS RN 139512-70-2
IUPAC Name:
4-chloro-5-fluorobenzene-1,2-diamine
Synonyms:
4-chloro-5-fluorobenzene-1,2-diamine
Molecular Weight (g/mol):
160.58
Molecular Formula:
C6H6ClFN2
InChi Key:
BSMPRJISGCTCDC-UHFFFAOYSA-N
SMILES:
NC1=CC(F)=C(Cl)C=C1N
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 4-Chloro-5-fluoro-o-phenylenediamine, 98%
CAS: 139512-70-2 Molecular Formula: C6H6ClFN2 Molecular Weight (g/mol): 160.58 MDL Number: MFCD00042485 InChI Key: BSMPRJISGCTCDC-UHFFFAOYSA-N PubChem CID: 735773 IUPAC Name: 4-chloro-5-fluorobenzene-1,2-diamine SMILES: NC1=CC(F)=C(Cl)C=C1N