CAS RN 140-66-9
CAS RN 140-66-9
IUPAC Name:
4-(2,4,4-trimethylpentan-2-yl)phenol
Synonyms:
4-tert-octylphenol p-terc.oktylfenol
Molecular Weight (g/mol):
206.33
Molecular Formula:
C14H22O
InChi Key:
ISAVYTVYFVQUDY-UHFFFAOYSA-N
SMILES:
CC(C)(C)CC(C)(C)C1=CC=C(O)C=C1