accessibility menu, dialog, popup

UserName

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 14003-34-0

H3CNNHO

CAS RN 14003-34-0

IUPAC Name: 3-methyl-1,2-dihydroquinoxalin-2-one
Synonyms: 3-methyl-1H-quinoxalin-2-one
Molecular Weight (g/mol): 160.18
Molecular Formula: C9H8N2O
InChi Key: BMIMNRPAEPIYDN-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=CC=C2NC1=O
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products