man

Test

CAS RN 14017-54-0

H2OH2OH2OH2OH2OH2OHoOClOOOOClOOOOClOOO

CAS RN 14017-54-0

IUPAC Name: holmium(3+) hexahydrate triperchlorate
Synonyms: holmium(3+) hexahydrate triperchlorate
Molecular Weight (g/mol): 571.36
Molecular Formula: Cl3H12HoO18
InChi Key: RRHAFXDVTMGQFK-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Ho+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results