accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 140191-40-8

NOOONOOONOOOY

CAS RN 140191-40-8

IUPAC Name: yttrium(3+) hydrate trinitrate
Synonyms: yttrium(3+) hydrate trinitrate
Molecular Weight (g/mol): 292.93
Molecular Formula: H2N3O10Y
InChi Key: WUVRZBFIXJWTGS-UHFFFAOYSA-N
SMILES: O.[Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Grade 
  • (4)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products