CAS RN 1403254-99-8
CAS RN 1403254-99-8
IUPAC Name:
N-[(4,6-dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl]-5-[ethyl(oxan-4-yl)amino]-4-methyl-4'-[(morpholin-4-yl)methyl]-[1,1'-biphenyl]-3-carboxamide
Synonyms:
N-[(4,6-dimethyl-2-oxo-1H-pyridin-3-yl)methyl]-5-[ethyl(oxan-4-yl)amino]-4-methyl-4'-(morpholin-4-ylmethyl)-[1,1'-biphenyl]-3-carboxamide
Molecular Weight (g/mol):
572.75
Molecular Formula:
C34H44N4O4
InChi Key:
NSQSAUGJQHDYNO-UHFFFAOYSA-N
SMILES:
CCN(C1CCOCC1)C1=CC(=CC(C(=O)NCC2=C(C)C=C(C)NC2=O)=C1C)C1=CC=C(CN2CCOCC2)C=C1