accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 14080-59-2

ClNNS

CAS RN 14080-59-2

IUPAC Name: 4-chlorothieno[2,3-d]pyrimidine
Synonyms: 4-chlorothieno[2,3-d]pyrimidine
Molecular Weight (g/mol): 170.61
Molecular Formula: C6H3ClN2S
InChi Key: NZCRUBBNZGVREM-UHFFFAOYSA-N
SMILES: ClC1=C2C=CSC2=NC=N1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products