CAS RN 1417997-93-3
CAS RN 1417997-93-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express HDAC8-IN-1, MedChemExpress
MedChemExpress HDAC8-IN-1 is a HDAC8 inhibitor with an IC50 of 27.2 nM.
