man

Test

CAS RN 1418308-27-6

H3CCH3NNONHNHOCH3H3C

CAS RN 1418308-27-6

IUPAC Name: 6-cyano-N-[(4,6-dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl]-1-(pentan-3-yl)-1H-indole-4-carboxamide
Synonyms: 6-cyano-N-[(4,6-dimethyl-2-oxo-1H-pyridin-3-yl)methyl]-1-(pentan-3-yl)indole-4-carboxamide
Molecular Weight (g/mol): 390.49
Molecular Formula: C23H26N4O2
InChi Key: PFHDWRIVDDIFRP-UHFFFAOYSA-N
SMILES: CCC(CC)N1C=CC2=C(C=C(C=C12)C#N)C(=O)NCC1=C(C)C=C(C)NC1=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results