accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 142-45-0

OHOHOO

CAS RN 142-45-0

IUPAC Name: but-2-ynedioic acid
Synonyms: acetylenedicarboxylic acid
Molecular Weight (g/mol): 114.06
Molecular Formula: C4H2O4
InChi Key: YTIVTFGABIZHHX-UHFFFAOYSA-N
SMILES: OC(=O)C#CC(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products