accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1421-65-4

CH3OO(S)NH2HOOHHCl

CAS RN 1421-65-4

IUPAC Name: hydrogen methyl 2-amino-3-(3,4-dihydroxyphenyl)propanoate chloride
Synonyms: hydrogen methyl 2-amino-3-(3,4-dihydroxyphenyl)propanoate chloride
Molecular Weight (g/mol): 247.68
Molecular Formula: C10H14ClNO4
InChi Key: WFGNJLMSYIJWII-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)C(N)CC1=CC=C(O)C(O)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products