CAS RN 1423186-80-4
CAS RN 1423186-80-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express GSK2983559 active metabolite, MedChemExpress
MedChemExpress GSK2983559 active metabolite is an active metabolite of GSK2983559. GSK2983559 active metabolite is a receptor interacting protein-2 (RIP2) kinase inhibitor extracted from patent WO/2014043446 A1, compound example 1.
