CAS RN 1427782-89-5
CAS RN 1427782-89-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express IWP L6, MedChemExpress
MedChemExpress IWP L6 (Porcn Inhibitor III) is a Porcn inhibitor with an EC50 of 0.5 nM.
