CAS RN 143128-30-7
CAS RN 143128-30-7
IUPAC Name:
1-[(4-iodophenyl)methyl]-1H-pyrazole
Synonyms:
1-[(4-iodophenyl)methyl]pyrazole
Molecular Weight (g/mol):
284.1
Molecular Formula:
C10H9IN2
InChi Key:
QRCIIHVBALOAQK-UHFFFAOYSA-N
SMILES:
IC1=CC=C(CN2C=CC=N2)C=C1