accessibility menu, dialog, popup

UserName

CAS RN 14323-36-5

PtNNNNKKH2OH2OH2O

CAS RN 14323-36-5

IUPAC Name: platinum(2+) dipotassium tetrakis(iminomethanide) tetrahydrate
Synonyms: platinum(2+) dipotassium tetrakis(cyanide) tetrahydrate
Molecular Weight (g/mol): 449.41
Molecular Formula: C4H8K2N4O4Pt
InChi Key: JJVBDLIBPBLHEW-UHFFFAOYSA-N
SMILES: O.O.O.O.[K+].[K+].[Pt++].[C-]#N.[C-]#N.[C-]#N.[C-]#N
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results