CAS RN 14328-52-0
CAS RN 14328-52-0
IUPAC Name:
2-amino-2-cyclohexylacetic acid
Synonyms:
cyclohexylglycine
Molecular Weight (g/mol):
157.21
Molecular Formula:
C8H15NO2
InChi Key:
WAMWSIDTKSNDCU-UHFFFAOYNA-N
SMILES:
NC(C1CCCCC1)C(O)=O
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar D-(-)-2-Cyclohexylglycine, 95%
CAS: 14328-52-0 Molecular Formula: C8H15NO2 Molecular Weight (g/mol): 157.213 MDL Number: MFCD01311678 InChI Key: WAMWSIDTKSNDCU-SSDOTTSWSA-N Synonym: d-cyclohexylglycine,d-alpha-cyclohexylglycine,r-2-amino-2-cyclohexylacetic acid,h-d-chg-oh,2-cyclohexyl-d-glycine,2r-amino cyclohexyl acetic acid,2r-2-amino-2-cyclohexylacetic acid,d-2-cyclohexylglycine,d-cyclohexyl glycine,h-cyclohexyl-d-gly-oh PubChem CID: 736849 IUPAC Name: (2R)-2-amino-2-cyclohexylacetic acid SMILES: C1CCC(CC1)C(C(=O)O)N