CAS RN 1438-82-0
CAS RN 1438-82-0
IUPAC Name:
dimethyl[(trimethylsilyl)oxy]silyl
Synonyms:
dimethyl[(trimethylsilyl)oxy]silyl
Molecular Weight (g/mol):
147.34
Molecular Formula:
C5H15OSi2
InChi Key:
XUKFPAQLGOOCNJ-UHFFFAOYSA-N
SMILES:
C[Si](C)O[Si](C)(C)C
1
–
1
of
1
results
1
Pentamethyldisiloxane, 95%, Thermo Scientific Chemicals
CAS: 1438-82-0 Molecular Formula: C5H15OSi2 Molecular Weight (g/mol): 147.344 MDL Number: MFCD00053658 InChI Key: XUKFPAQLGOOCNJ-UHFFFAOYSA-N Synonym: pentamethyldisiloxane,disiloxane, pentamethyl,1,1,1,3,3-pentamethyldisiloxane,dimethyl trimethylsilyloxy silicon,disiloxane, 1,1,1,3,3-pentamethyl,dimethyl trimethylsilyl oxy silyl,dimethylsilyl oxy trimethylsilane PubChem CID: 6327260 IUPAC Name: dimethyl(trimethylsilyloxy)silicon SMILES: C[Si](C)O[Si](C)(C)C