accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 144-83-2

NH2SOONHN

CAS RN 144-83-2

IUPAC Name: 4-amino-N-(pyridin-2-yl)benzene-1-sulfonamide
Synonyms: sulfapyridine sulfapyridine (substance) haptocil dagenan sulfapyridinum sulphapyridin trianon plurazol streptosilpyridine sulphapyridine
Molecular Weight (g/mol): 249.29
Molecular Formula: C11H11N3O2S
InChi Key: GECHUMIMRBOMGK-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products