CAS RN 14443-37-9
CAS RN 14443-37-9
IUPAC Name:
2-(methylsulfanyl)-6,7,8,9-tetrahydro-3H-purine-6,8-dione
Synonyms:
2-(methylsulfanyl)-7,9-dihydro-3H-purine-6,8-dione
Molecular Weight (g/mol):
198.2
Molecular Formula:
C6H6N4O2S
InChi Key:
ABNKHLJAVYVWBF-UHFFFAOYSA-N
SMILES:
CSC1=NC(=O)C2=C(NC(=O)N2)N1
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar 6,8-Dihydroxy-2-(methylthio)purine, 98%, Thermo Scientific™
CAS: 14443-37-9 Molecular Formula: C6H6N4O2S Molecular Weight (g/mol): 198.20 MDL Number: MFCD00022637 InChI Key: ABNKHLJAVYVWBF-UHFFFAOYSA-N Synonym: 6,8-dihydroxy-2-methylmercaptopurine,2-methylsulfanyl-7,9-dihydro-3h-purine-6,8-dione,2-methylthiopurine-6,8-diol,2-methylsulfanyl-7h-purine-6,8-diol,acmc-20am0i,6,8-dihydroxy-2-methylthio purine,2-methylthio-7h-purine-6,8-diol,6,8-dihydroxy-2-methylmercapto purine,2-methylthio-1h-purine-6,8 7h,9h-dione PubChem CID: 229271 SMILES: CSC1=NC(=O)C2=C(NC(=O)N2)N1