man

Test

CAS RN 1448317-68-7

CH3OOHSFFFFF

CAS RN 1448317-68-7

IUPAC Name: 3-methyl-5-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Synonyms: 3-methyl-5-(pentafluoro-λ⁶-sulfanyl)benzoic acid
Molecular Weight (g/mol): 262.19
Molecular Formula: C8H7F5O2S
InChi Key: DHRPCCNJMXXYMQ-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(=C1)C(O)=O)S(F)(F)(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results