accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1449135-43-6

BOOH3CCH3H3CH3CSONONOCH3CH3

CAS RN 1449135-43-6

IUPAC Name: 1,3-dimethyl-5-{[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methylidene}-1,3-diazinane-2,4,6-trione
Synonyms: 1,3-dimethyl-5-{[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophen-2-yl]methylidene}-1,3-diazinane-2,4,6-trione
Molecular Weight (g/mol): 376.23
Molecular Formula: C17H21BN2O5S
InChi Key: ACWASRSLONUFLW-UHFFFAOYSA-N
SMILES: CN1C(=O)N(C)C(=O)C(=CC2=CC=C(S2)B2OC(C)(C)C(C)(C)O2)C1=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products