accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 14590-13-7

NH4CoOPOOO

CAS RN 14590-13-7

IUPAC Name: λ²-cobalt(2+) ammonium phosphate
Synonyms: λ²-cobalt(2+) ammonium phosphate
Molecular Weight (g/mol): 171.94
Molecular Formula: CoH4NO4P
InChi Key: FCXHHGTWMFGYPP-UHFFFAOYSA-L
SMILES: [NH4+].[Co++].[O-]P([O-])([O-])=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products