accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 14667-55-1

CH3NCH3H3CN

CAS RN 14667-55-1

IUPAC Name: 2,3,5-trimethylpyrazine
Synonyms: trimethylpyrazine pyrazine, trimethyl-
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: IAEGWXHKWJGQAZ-UHFFFAOYSA-N
SMILES: CC1=CN=C(C)C(C)=N1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products