CAS RN 14691-88-4
CAS RN 14691-88-4
IUPAC Name:
1-hydroxy-2,2,6,6-tetramethylpiperidin-4-aminium
Synonyms:
1-hydroxy-2,2,6,6-tetramethylpiperidin-4-aminium
Molecular Weight (g/mol):
173.28
Molecular Formula:
C9H21N2O
InChi Key:
AYSOUYKQIUYGFR-UHFFFAOYSA-O
SMILES:
CC1(C)CC([NH3+])CC(C)(C)N1O
1
–
2
of
2
results
1
4-Amino-2,2,6,6-tetramethylpiperidinooxy, 97%, free radical, Thermo Scientific Chemicals
CAS: 14691-88-4 Molecular Formula: C9H19N2O Molecular Weight (g/mol): 171.26 MDL Number: MFCD00006479 Synonym: 4-Amino-TEMPO