man

Test

CAS RN 14692-17-2

OClOOOOClOOOOClOOODy

CAS RN 14692-17-2

IUPAC Name: dysprosium(3+) triperchlorate
Synonyms: dysprosium(3+) triperchlorate
Molecular Weight (g/mol): 460.84
Molecular Formula: Cl3DyO12
InChi Key: QNDHOIIJUAAORW-UHFFFAOYSA-K
SMILES: [Dy+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results