CAS RN 146982-24-3
CAS RN 146982-24-3
IUPAC Name:
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-oxo-4-(prop-2-en-1-yloxy)butanoic acid
Synonyms:
2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-oxo-4-(prop-2-en-1-yloxy)butanoic acid
Molecular Weight (g/mol):
395.41
Molecular Formula:
C22H21NO6
InChi Key:
FBNFRRNBFASDKS-UHFFFAOYNA-N
SMILES:
OC(=O)C(CC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1
–
1
of
1
results
1
N-Fmoc-L-aspartic acid 4-allyl ester, 98%, Thermo Scientific Chemicals
CAS: 146982-24-3 Molecular Formula: C22H21NO6 Molecular Weight (g/mol): 395.411 MDL Number: MFCD00190874 InChI Key: FBNFRRNBFASDKS-IBGZPJMESA-N Synonym: fmoc-asp oall-oh,fmoc-l-aspartic acid 4-allyl ester,fmoc-asp oallyl-oh,fmoc-asp oall oh,fmoc-asp-allyl ester,ambotzfaa1353,fmoc-asp oal-oh,pubchem12449,n-fmoc-l-aspartic acid 4-allyl ester,fmoc-asp oall-oh hplc PubChem CID: 10715692 IUPAC Name: (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-prop-2-enoxybutanoic acid SMILES: C=CCOC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13