accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 146982-27-6

H2COONH(S)OHONHOO

CAS RN 146982-27-6

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(prop-2-en-1-yloxy)carbonyl]amino}hexanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-6-{[(prop-2-en-1-yloxy)carbonyl]amino}hexanoic acid
Molecular Weight (g/mol): 452.51
Molecular Formula: C25H28N2O6
InChi Key: OJBNDXHENJDCBA-UHFFFAOYNA-N
SMILES: OC(=O)C(CCCCNC(=O)OCC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products