accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 147859-80-1

CH3HNOOOHNCH3CH3ONOOCH3

CAS RN 147859-80-1

IUPAC Name: methyl 1-(3-methyl-2-{[3-(propylcarbamoyl)oxiran-2-yl]formamido}pentanoyl)pyrrolidine-2-carboxylate
Synonyms: methyl 1-(3-methyl-2-{[3-(propylcarbamoyl)oxiran-2-yl]formamido}pentanoyl)pyrrolidine-2-carboxylate
Molecular Weight (g/mol): 397.47
Molecular Formula: C19H31N3O6
InChi Key: XGWSRLSPWIEMLQ-UHFFFAOYNA-N
SMILES: CCCNC(=O)C1OC1C(=O)NC(C(C)CC)C(=O)N1CCCC1C(=O)OC
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products