accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 148437-99-4

CH3OOOFFF

CAS RN 148437-99-4

IUPAC Name: methyl 2-(trifluoromethoxy)benzoate
Synonyms: methyl 2-(trifluoromethoxy)benzoate
Molecular Weight (g/mol): 220.15
Molecular Formula: C9H7F3O3
InChi Key: VDJYJSUDISVNRS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1OC(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products