accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 148546-82-1

BNCH3HOOH

CAS RN 148546-82-1

IUPAC Name: (4-methylpyridin-3-yl)boronic acid
Synonyms: 4-methylpyridin-3-ylboronic acid
Molecular Weight (g/mol): 136.95
Molecular Formula: C6H8BNO2
InChi Key: ASXFMIDIRZPCGK-UHFFFAOYSA-N
SMILES: CC1=C(C=NC=C1)B(O)O
1x1 image pixel for data collection 1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products