accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 149105-11-3

H3COOHFFF

CAS RN 149105-11-3

IUPAC Name: 1-[4-hydroxy-3-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[4-hydroxy-3-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: HKRUXZJKSFFSGF-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(=C(O)C=C1)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products