man

Test

CAS RN 150529-93-4

CH3H3CNONO

CAS RN 150529-93-4

IUPAC Name: 4-phenyl-2-[2-(4-phenyl-4,5-dihydro-1,3-oxazol-2-yl)propan-2-yl]-4,5-dihydro-1,3-oxazole
Synonyms: 4-phenyl-2-[2-(4-phenyl-4,5-dihydro-1,3-oxazol-2-yl)propan-2-yl]-4,5-dihydro-1,3-oxazole
Molecular Weight (g/mol): 334.42
Molecular Formula: C21H22N2O2
InChi Key: JTNVCJCSECAMLD-UHFFFAOYNA-N
SMILES: CC(C)(C1=NC(CO1)C1=CC=CC=C1)C1=NC(CO1)C1=CC=CC=C1
Melting Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results