CAS RN 150780-67-9
CAS RN 150780-67-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific™ 2'-OMe-iBu-G Phosphoramidite, standard grade, 20-400 finish bottle
2'-OMe-iBu-G CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with 2'-O-methyl protection, for use in oligonucleotide synthesis.