man

Test

CAS RN 15118-67-9

H3COOOOCH3

CAS RN 15118-67-9

IUPAC Name: ethyl 4-(4-methoxyphenyl)-4-oxobutanoate
Synonyms: ethyl 4-(4-methoxyphenyl)-4-oxobutanoate
Molecular Weight (g/mol): 236.27
Molecular Formula: C13H16O4
InChi Key: FYUAOZFEVHSJTO-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC(=O)C1=CC=C(OC)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results