CAS RN 15126-06-4
CAS RN 15126-06-4
IUPAC Name:
methyl 4-hydroxy-3-iodobenzoate
Synonyms:
methyl 4-hydroxy-3-iodobenzoate
Molecular Weight (g/mol):
278.05
Molecular Formula:
C8H7IO3
InChi Key:
PXNOLLHARLSLHY-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(I)=C(O)C=C1