accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 15128-82-2

OHNNOO

CAS RN 15128-82-2

IUPAC Name: 2-nitropyridin-3-ol
Synonyms: 3-pyridinol, 2-nitro-
Molecular Weight (g/mol): 140.1
Molecular Formula: C5H4N2O3
InChi Key: QBPDSKPWYWIHGA-UHFFFAOYSA-N
SMILES: OC1=CC=CN=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products