CAS RN 15280-57-6
CAS RN 15280-57-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Thermo Scientific Alfa Aesar Erbium(III) acetate tetrahydrate, 99% (REO)
CAS: 15280-57-6 Molecular Formula: C6H9ErO6·4H2O MDL Number: MFCD00013034
2
Thermo Scientific Alfa Aesar Erbium(III) acetate tetrahydrate, REacton™, 99.9% (REO)
CAS: 15280-57-6 MDL Number: MFCD00013034