accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 15337-84-5

ClClPtH3CSH3CCH3SCH3

CAS RN 15337-84-5

IUPAC Name: platinum(2+) bis((ethylsulfanyl)ethane) dichloride
Synonyms: platinum(2+) bis(ethyl sulfide) dichloride
Molecular Weight (g/mol): 446.35
Molecular Formula: C8H20Cl2PtS2
InChi Key: DCIVLRIRJUWCIS-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pt++].CCSCC.CCSCC
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products