accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 155141-29-0

CH3NOSOHNONOHOOHO(Z)

CAS RN 155141-29-0

IUPAC Name: (2Z)-but-2-enedioic acid; 5-[(4-{2-[methyl(pyridin-2-yl)amino]ethoxy}phenyl)methyl]-1,3-thiazolidine-2,4-dione
Synonyms: maleic acid; rosiglitazone
Molecular Weight (g/mol): 473.5
Molecular Formula: C22H23N3O7S
InChi Key: SUFUKZSWUHZXAV-BTJKTKAUNA-N
SMILES: OC(=O)\C=C/C(O)=O.CN(CCOC1=CC=C(CC2SC(=O)NC2=O)C=C1)C1=CC=CC=N1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products