accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1552-92-7

CH3OO

CAS RN 1552-92-7

IUPAC Name: ethyl 2-cyclohexylideneacetate
Synonyms: ethyl 2-cyclohexylideneacetate
Molecular Weight (g/mol): 168.24
Molecular Formula: C10H16O2
InChi Key: MCWDXHYYYNGYGK-UHFFFAOYSA-N
SMILES: CCOC(=O)C=C1CCCCC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products