CAS RN 15525-64-1
CAS RN 15525-64-1
IUPAC Name:
silver(1+) 2,4-dioxopentan-3-ide
Synonyms:
silver(1+) acetylacetonate
Molecular Weight (g/mol):
206.98
Molecular Formula:
C5H7AgO2
InChi Key:
UEGHNEVWUGTMES-UHFFFAOYSA-N
SMILES:
[Ag+].CC(=O)[CH-]C(C)=O
1
–
1
of
1
results
1
Silver 2,4-pentanedionate, 98%, Thermo Scientific Chemicals
CAS: 15525-64-1 Molecular Formula: C5H8AgO2 Molecular Weight (g/mol): 207.985 MDL Number: MFCD00000012 InChI Key: LEUOXKLUFCTIIY-LNKPDPKZSA-N Synonym: silver acetylacetonate,2,4-pentanedione, silver derivative,silver, 2,4-pentanedionato-o,o',silver 2,4-pentanedionate,4-oxopent-2-en-2-yl oxy silver,3z-4-argentiooxy pent-3-en-2-one PubChem CID: 11074572 IUPAC Name: (Z)-4-hydroxypent-3-en-2-one;silver SMILES: CC(=CC(=O)C)O.[Ag]