accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 15570-12-4

H3COHS

CAS RN 15570-12-4

IUPAC Name: 3-methoxybenzene-1-thiol
Synonyms: 3-methoxy thiophenol
Molecular Weight (g/mol): 140.2
Molecular Formula: C7H8OS
InChi Key: QMVAZEHZOPDGHA-UHFFFAOYSA-N
SMILES: COC1=CC=CC(S)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products