man

Test

CAS RN 15662-33-6

H3CCH3OHOOHNHOCH3HOOHOCH3OHOHH3C

CAS RN 15662-33-6

IUPAC Name: (1R,2R,3S,6S,7S,9S,10S,11S,12R,13S,14R)-2,6,9,11,13,14-hexahydroxy-3,7,10-trimethyl-11-(propan-2-yl)-15-oxapentacyclo[7.5.1.0¹,⁶.0⁷,¹³.0¹⁰,¹⁴]pentadecan-12-yl 1H-pyrrole-2-carboxylate
Synonyms: ryania bonide ryatox ground ryania specisa(vahl) stemwood (alkoloid ryanodine) ryania speciosa ryanodine (substance) ryanexel ryania powder ryanicide
Molecular Weight (g/mol): 493.55
Molecular Formula: C25H35NO9
InChi Key: JJSYXNQGLHBRRK-YSOSZROBSA-N
SMILES: CC(C)[C@@]1(O)[C@@H](OC(=O)C2=CC=CN2)[C@@]2(O)[C@@]3(C)C[C@]4(O)O[C@@]5([C@H](O)[C@@H](C)CC[C@]35O)[C@@]2(O)[C@]14C

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results